Recombinat Protein,Human CD99 MIC2 Protein, His Tag,Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human CD99 MIC2 Protein, His Tag,Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human CD99 MIC2 Protein, His Tag,Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 1 mg ACRBCD9-H5221-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD9-H5221-1MG
Lokale Artikelnummer:: ACRBCD9-H5221-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder