Biotinylated Human CD16a (V176),Avitag,His Tag (SPR BLI verified) (CDA-H82E9) is expressed from human 293 cells (HEK293). It conta ins AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human CD16a (V176),Avitag,His Tag (SPR BLI verified) (CDA-H82E9) is expressed from human 293 cells (HEK293). It conta ins AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human CD16a (V176), Avitag,His Tag (SPR  BLI verified) (CDA-H82E9) is expressed from human 293 cells (HEK293). It conta ins AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 200 µG ACRBCDA-H82E9200UG 1 * 200 µG
Hersteller-Artikelnummer:: CDA-H82E9200UG
Lokale Artikelnummer:: ACRBCDA-H82E9200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder