Biotinylated Human CD32b, Avitag,His Tag (SPR BLI verified) (CDB-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession P31994-1). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human CD32b, Avitag,His Tag (SPR BLI verified) (CDB-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession P31994-1). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human CD32b, Avitag,His Tag (SPR  BLI verified) (CDB-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession P31994-1). 1 * 25 µG ACRBCDB-H82E0-25UG 1 * 25 µG
Hersteller-Artikelnummer:: CDB-H82E0-25UG
Lokale Artikelnummer:: ACRBCDB-H82E0-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder