Biotinylated Cynomolgus CD3ECD3D Heterodimer Protein, His,AvitagTag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) Phe 22 - Ala 105 (CD3D) (Accession Q95LI5-1 (CD3E) Q95LI8-1 (CD3D)). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Cynomolgus CD3ECD3D Heterodimer Protein, His,AvitagTag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) Phe 22 - Ala 105 (CD3D) (Accession Q95LI5-1 (CD3E) Q95LI8-1 (CD3D)). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Cynomolgus CD3ECD3D Heterodimer Protein, His,AvitagTag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) Phe 22 - Ala 105 (CD3D) (Accession Q95LI5-1 (CD3E) Q95LI8-1 (CD3D)). 1 * 200 µG ACRBCDD-C82W6200UG 1 * 200 µG
Hersteller-Artikelnummer:: CDD-C82W6200UG
Lokale Artikelnummer:: ACRBCDD-C82W6200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder