Cynomolgus CD3 epsilon, Fc,His Tag (CDE-C5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accessio n Q95LI5). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Cynomolgus CD3 epsilon, Fc,His Tag (CDE-C5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accessio n Q95LI5). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Cynomolgus CD3 epsilon, Fc,His Tag (CDE-C5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accessio n Q95LI5). 1 * 100 µG ACRBCDE-C5254100UG 1 * 100 µG
Hersteller-Artikelnummer:: CDE-C5254100UG
Lokale Artikelnummer:: ACRBCDE-C5254100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder