Synonym FLJ18683, T3E, TCRE, CD3E, CD3-epsilon Source HRP-Human CD3 epsilon, His Tag(CDE-HR2H4) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # P07766-1). Predicted N-terminus: Asp 23 Formulation Lyophilized f 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Synonym FLJ18683, T3E, TCRE, CD3E, CD3-epsilon Source HRP-Human CD3 epsilon, His Tag(CDE-HR2H4) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # P07766-1). Predicted N-terminus: Asp 23 Formulation Lyophilized f 1 * 25 G
PRODUKT
Erweitert / Kompakt
Synonym FLJ18683, T3E, TCRE, CD3E, CD3-epsilon Source HRP-Human CD3 epsilon, His Tag(CDE-HR2H4) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # P07766-1). Predicted N-terminus: Asp 23 Formulation Lyophilized f 1 * 25 µG ACRBCDE-HR2H4-25UG 1 * 25 µG
Hersteller-Artikelnummer:: CDE-HR2H4-25UG
Lokale Artikelnummer:: ACRBCDE-HR2H4-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder