Human Rhesus macaque CD40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H5256) is expressed from human 293 cells (HEK293). It cont ains AA Met 113 - Leu 261 (Accession P29965-1). In the region Met 113 - Leu 261, the AA sequence of Human and Rhesus L 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human Rhesus macaque CD40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H5256) is expressed from human 293 cells (HEK293). It cont ains AA Met 113 - Leu 261 (Accession P29965-1). In the region Met 113 - Leu 261, the AA sequence of Human and Rhesus L 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human Rhesus macaque CD40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H5256) is expressed from human 293 cells (HEK293). It cont ains AA Met 113 - Leu 261 (Accession P29965-1). In the region Met 113 - Leu 261, the AA sequence of Human and Rhesus L 1 * 1 mg ACRBCDL-H5256-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CDL-H5256-1MG
Lokale Artikelnummer:: ACRBCDL-H5256-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder