Human CD20 Full Length Protein-VLP (CDP-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human CD20 Full Length Protein-VLP (CDP-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human CD20 Full Length Protein-VLP (CDP-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession 1 * 100 µG ACRBCDP-H52P6100UG 1 * 100 µG
Hersteller-Artikelnummer:: CDP-H52P6100UG
Lokale Artikelnummer:: ACRBCDP-H52P6100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder