Human Complement C5 (R885H), His Tag (CO5-H52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (R885H)). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human Complement C5 (R885H), His Tag (CO5-H52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (R885H)). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human Complement C5 (R885H), His Tag (CO5-H52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (R885H)). 1 * 1 mg ACRBCO5-H52HX-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CO5-H52HX-1MG
Lokale Artikelnummer:: ACRBCO5-H52HX-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder