Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession P0 6684-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession P0 6684-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession P0 6684-1). 1 * 100 µG ACRBCO5-M52H4100UG 1 * 100 µG
Hersteller-Artikelnummer:: CO5-M52H4100UG
Lokale Artikelnummer:: ACRBCO5-M52H4100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder