Biotinylated Mouse M-CSF R, Avitag,His Tag (CSR-M82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 ( Accession P09581-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Mouse M-CSF R, Avitag,His Tag (CSR-M82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 ( Accession P09581-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Mouse M-CSF R, Avitag,His Tag (CSR-M82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 ( Accession P09581-1). 1 * 200 µG ACRBCSR-M82E8200UG 1 * 200 µG
Hersteller-Artikelnummer:: CSR-M82E8200UG
Lokale Artikelnummer:: ACRBCSR-M82E8200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder