Cynomolgus Rhesus macaque CTLA-4, His Tag (CT4-C5227) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 ( Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homo 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque CTLA-4, His Tag (CT4-C5227) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 ( Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homo 1 * 200 G
PRODUKT
Erweitert / Kompakt
Cynomolgus Rhesus macaque CTLA-4, His Tag (CT4-C5227) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 ( Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homo 1 * 200 µG ACRBCT4-C5227200UG 1 * 200 µG
Hersteller-Artikelnummer:: CT4-C5227200UG
Lokale Artikelnummer:: ACRBCT4-C5227200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder