Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession O54908). 1 * 500 G
Lieferant: ACROBIOSYSTEMS
Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession O54908). 1 * 500 G
PRODUKT
Erweitert / Kompakt
Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession O54908). 1 * 500 µG ACRBDK1-M52H6500UG 1 * 500 µG
Hersteller-Artikelnummer:: DK1-M52H6500UG
Lokale Artikelnummer:: ACRBDK1-M52H6500UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder