Recombinat Protein,Mouse Dkk-3 Protein, His Tag,Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It cont ains AA Pro 23 - Ile 349 (Accession NP056629.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 50 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Mouse Dkk-3 Protein, His Tag,Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It cont ains AA Pro 23 - Ile 349 (Accession NP056629.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 50 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Mouse Dkk-3 Protein, His Tag,Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It cont ains AA Pro 23 - Ile 349 (Accession NP056629.1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 50 µG ACRBDK3-M5226-50UG 1 * 50 µG
Hersteller-Artikelnummer:: DK3-M5226-50UG
Lokale Artikelnummer:: ACRBDK3-M5226-50UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder