Recombinat Protein,Human DLL1 Delta1 Protein, His Tag,Human DLL1, His Tag (DL1-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Gln 18 - Gly 540 (Accession NP005609).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human DLL1 Delta1 Protein, His Tag,Human DLL1, His Tag (DL1-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Gln 18 - Gly 540 (Accession NP005609).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human DLL1 Delta1 Protein, His Tag,Human DLL1, His Tag (DL1-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Gln 18 - Gly 540 (Accession NP005609).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg ACRBDL1-H52H8-1MG 1 * 1 mg
Hersteller-Artikelnummer:: DL1-H52H8-1MG
Lokale Artikelnummer:: ACRBDL1-H52H8-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder