Biotinylated Human DNAM-1, Fc,Avitag (DN1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Access ion NP006557). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human DNAM-1, Fc,Avitag (DN1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Access ion NP006557). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human DNAM-1, Fc,Avitag (DN1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Access ion NP006557). 1 * 25 µG ACRBDN1-H82F9-25UG 1 * 25 µG
Hersteller-Artikelnummer:: DN1-H82F9-25UG
Lokale Artikelnummer:: ACRBDN1-H82F9-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder