Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW he 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW he 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW he 1 * 1 mg ACRBDP4-H5211-1MG 1 * 1 mg
Hersteller-Artikelnummer:: DP4-H5211-1MG
Lokale Artikelnummer:: ACRBDP4-H5211-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder