Recombinat Protein,Human DPPIV CD26 Protein, Fc Tag,Human DPPIV, Fc Tag (DP4-H5266) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries a human IgG1 Fc tag at the N-terminus. The pr 1 * 50 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human DPPIV CD26 Protein, Fc Tag,Human DPPIV, Fc Tag (DP4-H5266) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries a human IgG1 Fc tag at the N-terminus. The pr 1 * 50 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human DPPIV CD26 Protein, Fc Tag,Human DPPIV, Fc Tag (DP4-H5266) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries a human IgG1 Fc tag at the N-terminus. The pr 1 * 50 µG ACRBDP4-H5266-50UG 1 * 50 µG
Hersteller-Artikelnummer:: DP4-H5266-50UG
Lokale Artikelnummer:: ACRBDP4-H5266-50UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder