Recombinat Protein,Human Ephrin-A4 EFNA4 Protein, His Tag,Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (H EK293). It contains AA Leu 26 - Gly 171 (Accession AAI07484).This protein carries a polyhistidine tag at the C-termin 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Ephrin-A4 EFNA4 Protein, His Tag,Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (H EK293). It contains AA Leu 26 - Gly 171 (Accession AAI07484).This protein carries a polyhistidine tag at the C-termin 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Ephrin-A4 EFNA4 Protein, His Tag,Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (H EK293). It contains AA Leu 26 - Gly 171 (Accession AAI07484).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG ACRBEA4-H5229100UG 1 * 100 µG
Hersteller-Artikelnummer:: EA4-H5229100UG
Lokale Artikelnummer:: ACRBEA4-H5229100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder