FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). 1 * 200 G
PRODUKT
Erweitert / Kompakt
FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). 1 * 200 µG ACRBEGR-HF2H5200UG 1 * 200 µG
Hersteller-Artikelnummer:: EGR-HF2H5200UG
Lokale Artikelnummer:: ACRBEGR-HF2H5200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder