Recombinat Protein,ActiveMax Human Erythropoietin EPO Protein, Tag Free,ActiveMax Human EPO, Tag Free (EPO-H4214) is expressed fr om human 293 cells (HEK293). It contains AA Ala 28 - Arg 193 (Accession AAH93628.1).This protein carries no tag. The c 1 * 20 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,ActiveMax Human Erythropoietin EPO Protein, Tag Free,ActiveMax Human EPO, Tag Free (EPO-H4214) is expressed fr om human 293 cells (HEK293). It contains AA Ala 28 - Arg 193 (Accession AAH93628.1).This protein carries no tag. The c 1 * 20 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,ActiveMax® Human Erythropoietin EPO Protein, Tag Free,ActiveMax® Human EPO, Tag Free (EPO-H4214) is expressed fr om human 293 cells (HEK293). It contains AA Ala 28 - Arg 193 (Accession AAH93628.1).This protein carries no tag. The c 1 * 20 µG ACRBEPO-H4214-20UG 1 * 20 µG
Hersteller-Artikelnummer:: EPO-H4214-20UG
Lokale Artikelnummer:: ACRBEPO-H4214-20UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder