Recombinat Protein,Human FABP2 I-FABP Protein, His Tag,Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession NP000125).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human FABP2 I-FABP Protein, His Tag,Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession NP000125).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human FABP2 I-FABP Protein, His Tag,Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession NP000125).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg ACRBFA2-H5149-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FA2-H5149-1MG
Lokale Artikelnummer:: ACRBFA2-H5149-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder