Recombinat Protein,Human FABP5 E-FABP Protein, His Tag,Human FABP5, His Tag (FA5-H5140) is expressed from E.coli cells. It contains AA Ala 2 - Glu 135 (Accession AAH19385).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human FABP5 E-FABP Protein, His Tag,Human FABP5, His Tag (FA5-H5140) is expressed from E.coli cells. It contains AA Ala 2 - Glu 135 (Accession AAH19385).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human FABP5 E-FABP Protein, His Tag,Human FABP5, His Tag (FA5-H5140) is expressed from E.coli cells. It contains AA Ala 2 - Glu 135 (Accession AAH19385).This protein carries a polyhistidine tag at the N-terminus. The protein has W 1 * 1 mg ACRBFA5-H5140-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FA5-H5140-1MG
Lokale Artikelnummer:: ACRBFA5-H5140-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder