Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It cont ains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region 97 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It cont ains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region 97 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It cont ains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region 97 1 * 1 mg ACRBFCM-C52W9-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FCM-C52W9-1MG
Lokale Artikelnummer:: ACRBFCM-C52W9-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder