Biotinylated Mouse FCGRTB2M Heterodimer Protein, His,Avitag (BLI verified) (FCM-M82W5) is expressed from human 293 cells (HEK293). I t contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q61559-1 (FCGRT) P01887-1 (B2M)). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Mouse FCGRTB2M Heterodimer Protein, His,Avitag (BLI verified) (FCM-M82W5) is expressed from human 293 cells (HEK293). I t contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q61559-1 (FCGRT) P01887-1 (B2M)). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Mouse FCGRTB2M Heterodimer Protein, His,Avitag (BLI verified) (FCM-M82W5) is expressed from human 293 cells (HEK293). I t contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession  Q61559-1 (FCGRT) P01887-1 (B2M)). 1 * 200 µG ACRBFCM-M82W5200UG 1 * 200 µG
Hersteller-Artikelnummer:: FCM-M82W5200UG
Lokale Artikelnummer:: ACRBFCM-M82W5200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder