Recombinat Protein,Human Fetuin A FETUA Protein, His Tag,Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK 293). It contains AA Ala 19 - Val 367 (Accession NP001613).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Fetuin A FETUA Protein, His Tag,Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK 293). It contains AA Ala 19 - Val 367 (Accession NP001613).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Fetuin A FETUA Protein, His Tag,Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK 293). It contains AA Ala 19 - Val 367 (Accession NP001613).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg ACRBFEA-H5221-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FEA-H5221-1MG
Lokale Artikelnummer:: ACRBFEA-H5221-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder