Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 100 µG ACRBFG1-H52HY100UG 1 * 100 µG
Hersteller-Artikelnummer:: FG1-H52HY100UG
Lokale Artikelnummer:: ACRBFG1-H52HY100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder