Recombinat Protein,Human FGF R4 CD334 Protein, His Tag,Human FGF R4, His Tag (FG4-H5228) is expressed from human 293 cells (HEK293) . It contains AA Leu 22 - Asp 369 (Accession P22455-1).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human FGF R4 CD334 Protein, His Tag,Human FGF R4, His Tag (FG4-H5228) is expressed from human 293 cells (HEK293) . It contains AA Leu 22 - Asp 369 (Accession P22455-1).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human FGF R4 CD334 Protein, His Tag,Human FGF R4, His Tag (FG4-H5228) is expressed from human 293 cells (HEK293) . It contains AA Leu 22 - Asp 369 (Accession P22455-1).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 1 mg ACRBFG4-H5228-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FG4-H5228-1MG
Lokale Artikelnummer:: ACRBFG4-H5228-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder