Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acc ession P35916-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acc ession P35916-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acc ession P35916-1). 1 * 200 µG ACRBFL4-H82E1200UG 1 * 200 µG
Hersteller-Artikelnummer:: FL4-H82E1200UG
Lokale Artikelnummer:: ACRBFL4-H82E1200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder