Biotinylated Mouse FOLR1, His,Avitag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Access ion P35846-1). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Mouse FOLR1, His,Avitag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Access ion P35846-1). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Mouse FOLR1, His,Avitag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Access ion P35846-1). 1 * 25 µG ACRBFO1-M82E9-25UG 1 * 25 µG
Hersteller-Artikelnummer:: FO1-M82E9-25UG
Lokale Artikelnummer:: ACRBFO1-M82E9-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder