Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HE K293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HE K293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HE K293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg ACRBFZ5-H5259-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FZ5-H5259-1MG
Lokale Artikelnummer:: ACRBFZ5-H5259-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder