Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg ACRBFZ7-H52H9-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FZ7-H52H9-1MG
Lokale Artikelnummer:: ACRBFZ7-H52H9-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder