Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH69 319.1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH69 319.1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH69 319.1). 1 * 100 µG ACRBGIL-H5249100UG 1 * 100 µG
Hersteller-Artikelnummer:: GIL-H5249100UG
Lokale Artikelnummer:: ACRBGIL-H5249100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder