Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (GP3-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP005594665.1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (GP3-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP005594665.1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (GP3-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP005594665.1). 1 * 1 mg ACRBGP3-C5259-1MG 1 * 1 mg
Hersteller-Artikelnummer:: GP3-C5259-1MG
Lokale Artikelnummer:: ACRBGP3-C5259-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder