Recombinat Protein,Human Osteoactivin GPNMB Protein, His Tag,Human Osteoactivin, His Tag (GPB-H5229) is expressed from human 293 ce lls (HEK293). It contains AA Ala 22 - Pro 486 (Accession AAH32783).This protein carries a polyhistidine tag at the C- 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Osteoactivin GPNMB Protein, His Tag,Human Osteoactivin, His Tag (GPB-H5229) is expressed from human 293 ce lls (HEK293). It contains AA Ala 22 - Pro 486 (Accession AAH32783).This protein carries a polyhistidine tag at the C- 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Osteoactivin GPNMB Protein, His Tag,Human Osteoactivin, His Tag (GPB-H5229) is expressed from human 293 ce lls (HEK293). It contains AA Ala 22 - Pro 486 (Accession AAH32783).This protein carries a polyhistidine tag at the C- 1 * 1 mg ACRBGPB-H5229-1MG 1 * 1 mg
Hersteller-Artikelnummer:: GPB-H5229-1MG
Lokale Artikelnummer:: ACRBGPB-H5229-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder