Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg ACRBGRA-H52H7-1MG 1 * 1 mg
Hersteller-Artikelnummer:: GRA-H52H7-1MG
Lokale Artikelnummer:: ACRBGRA-H52H7-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder