Recombinat Protein,Human HE4 WFDC2 Protein, His Tag,Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It c ontains AA Glu 31 - Phe 124 (Accession AAH46106).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human HE4 WFDC2 Protein, His Tag,Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It c ontains AA Glu 31 - Phe 124 (Accession AAH46106).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human HE4 WFDC2 Protein, His Tag,Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It c ontains AA Glu 31 - Phe 124 (Accession AAH46106).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 100 µG ACRBHE4-H5223100UG 1 * 100 µG
Hersteller-Artikelnummer:: HE4-H5223100UG
Lokale Artikelnummer:: ACRBHE4-H5223100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder