Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 1 mg ACRBHST-H5245-1MG 1 * 1 mg
Hersteller-Artikelnummer:: HST-H5245-1MG
Lokale Artikelnummer:: ACRBHST-H5245-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder