Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH05 333). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH05 333). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH05 333). 1 * 200 µG ACRBIF1-H5254200UG 1 * 200 µG
Hersteller-Artikelnummer:: IF1-H5254200UG
Lokale Artikelnummer:: ACRBIF1-H5254200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder