Human IGF-II, Fc Tag (IG2-H4260) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession P01344-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human IGF-II, Fc Tag (IG2-H4260) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession P01344-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human IGF-II, Fc Tag (IG2-H4260) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession P01344-1). 1 * 100 µG ACRBIG2-H4260100UG 1 * 100 µG
Hersteller-Artikelnummer:: IG2-H4260100UG
Lokale Artikelnummer:: ACRBIG2-H4260100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder