Human IL-21, Fc Tag (IL1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accession Q9HBE4-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human IL-21, Fc Tag (IL1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accession Q9HBE4-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human IL-21, Fc Tag (IL1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accession Q9HBE4-1). 1 * 100 µG ACRBIL1-H5253100UG 1 * 100 µG
Hersteller-Artikelnummer:: IL1-H5253100UG
Lokale Artikelnummer:: ACRBIL1-H5253100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder