Mouse IL-13RA1, Fc Tag (IL1-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession NP598751) . 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Mouse IL-13RA1, Fc Tag (IL1-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession NP598751) . 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Mouse IL-13RA1, Fc Tag (IL1-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession NP598751) . 1 * 1 mg ACRBIL1-M5258-1MG 1 * 1 mg
Hersteller-Artikelnummer:: IL1-M5258-1MG
Lokale Artikelnummer:: ACRBIL1-M5258-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder