Human IL-1 RII, Fc Tag (IL2-H4256) is expressed from human 293 cells (HEK293). It contains AA Phe 14 - Glu 343 (Accession NP004624. 1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human IL-1 RII, Fc Tag (IL2-H4256) is expressed from human 293 cells (HEK293). It contains AA Phe 14 - Glu 343 (Accession NP004624. 1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human IL-1 RII, Fc Tag (IL2-H4256) is expressed from human 293 cells (HEK293). It contains AA Phe 14 - Glu 343 (Accession NP004624. 1). 1 * 1 mg ACRBIL2-H4256-1MG 1 * 1 mg
Hersteller-Artikelnummer:: IL2-H4256-1MG
Lokale Artikelnummer:: ACRBIL2-H4256-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder