Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 1 mg ACRBIL3-H52H7-1MG 1 * 1 mg
Hersteller-Artikelnummer:: IL3-H52H7-1MG
Lokale Artikelnummer:: ACRBIL3-H52H7-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder