Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accessi on P40933-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accessi on P40933-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accessi on P40933-1). 1 * 200 µG ACRBIL5-H82F3200UG 1 * 200 µG
Hersteller-Artikelnummer:: IL5-H82F3200UG
Lokale Artikelnummer:: ACRBIL5-H82F3200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder