Recombinat Protein,Human IL-3 R alpha CD123 Protein, Llama IgG2b Fc Tag, low endotoxin,Human IL-3 R alpha, Llama IgG2b Fc Tag, low endotoxin (ILA-H5255) is expressed from human 293 cells (HEK293). It contains AA Thr 19-Arg 305 (Accession P26951-1 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human IL-3 R alpha CD123 Protein, Llama IgG2b Fc Tag, low endotoxin,Human IL-3 R alpha, Llama IgG2b Fc Tag, low endotoxin (ILA-H5255) is expressed from human 293 cells (HEK293). It contains AA Thr 19-Arg 305 (Accession P26951-1 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human IL-3 R alpha CD123 Protein, Llama IgG2b Fc Tag, low endotoxin,Human IL-3 R alpha, Llama IgG2b Fc Tag, low endotoxin (ILA-H5255) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305  (Accession P26951-1 1 * 1 mg ACRBILA-H5255-1MG 1 * 1 mg
Hersteller-Artikelnummer:: ILA-H5255-1MG
Lokale Artikelnummer:: ACRBILA-H5255-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder