Recombinat Protein,Human IL-37 IL-1F7b Protein, His Tag,Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contain s AA Val 46 - Asp 218 (Accession AAH20637).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 500 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human IL-37 IL-1F7b Protein, His Tag,Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contain s AA Val 46 - Asp 218 (Accession AAH20637).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 500 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human IL-37 IL-1F7b Protein, His Tag,Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contain s AA Val 46 - Asp 218 (Accession AAH20637).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 500 µG ACRBILB-H5141500UG 1 * 500 µG
Hersteller-Artikelnummer:: ILB-H5141500UG
Lokale Artikelnummer:: ACRBILB-H5141500UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder