Biotinylated Human IL-17AIL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) Arg 31 - Gln 163 (IL-17F) (Accession Q16552-1 (IL-17A) Q96PD4-1 (IL-17F)). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human IL-17AIL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) Arg 31 - Gln 163 (IL-17F) (Accession Q16552-1 (IL-17A) Q96PD4-1 (IL-17F)). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human IL-17AIL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) Arg 31 - Gln 163 (IL-17F) (Accession Q16552-1 (IL-17A) Q96PD4-1 (IL-17F)). 1 * 200 µG ACRBILF-H82W1200UG 1 * 200 µG
Hersteller-Artikelnummer:: ILF-H82W1200UG
Lokale Artikelnummer:: ACRBILF-H82W1200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder