Recombinat Protein,Human IL-18BP Protein, Fc Tag,Human IL-18BP, Fc Tag (ILP-H5253) is expressed from human 293 cells (HEK293). It co ntains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a human IgG1 Fc tag at the C-terminus. The p 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human IL-18BP Protein, Fc Tag,Human IL-18BP, Fc Tag (ILP-H5253) is expressed from human 293 cells (HEK293). It co ntains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a human IgG1 Fc tag at the C-terminus. The p 1 * 200 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human IL-18BP Protein, Fc Tag,Human IL-18BP, Fc Tag (ILP-H5253) is expressed from human 293 cells (HEK293). It co ntains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a human IgG1 Fc tag at the C-terminus. The p 1 * 200 µG ACRBILP-H5253200UG 1 * 200 µG
Hersteller-Artikelnummer:: ILP-H5253200UG
Lokale Artikelnummer:: ACRBILP-H5253200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder