Human IL-21 R, His Tag (ILR-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession AAH04348) . 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Human IL-21 R, His Tag (ILR-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession AAH04348) . 1 * 200 G
PRODUKT
Erweitert / Kompakt
Human IL-21 R, His Tag (ILR-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession AAH04348) . 1 * 200 µG ACRBILR-H5226200UG 1 * 200 µG
Hersteller-Artikelnummer:: ILR-H5226200UG
Lokale Artikelnummer:: ACRBILR-H5226200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder